Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefprozil

Chemical Identifiers

Chemical Name or Material Cefprozil
CAS 92665-29-7
Formula Weight 389.42
MDL Number MFCD00865082
Merck Index 14,1942
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6632603

  SDS
alfa aesar™
J6632603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66326MC

  SDS
alfa aesar™
J66326MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 389.42
Melting Point 218° to 225°C
Chemical Name or Material Cefprozil
MDL Number MFCD00865082
CAS 92665-29-7
Boiling Point 803°C
Storage Note 1 2° to 8°C
RTECS Number XI0339000
Physical Form Powder
Merck Index 14,1942
Solubility Information Soluble in DMSO at 10mg/mL; Insoluble in water
Color White
Quantity 1g
Safety and Handling

2° to 8°C