Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefpodoxime proxetil, 96%

A broad spectrum antibiotic

Chemical Identifiers

Chemical Name or Material Cefpodoxime proxetil
CAS 87239-81-4
Formula Weight 557.6
MDL Number MFCD00865088
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6606103

 
alfa aesar™
J6606103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 111° to 113°C
Formula Weight 557.6
Chemical Name or Material Cefpodoxime proxetil
MDL Number MFCD00865088
CAS 87239-81-4
Assay 96%
Physical Form Powder
Storage Note 1 2° to 8°C
Solubility Information Soluble in DMSO
Quantity 1g
RTECS Number XI0367370
Sensitivity Light Sensitive
Color White to off-white
Safety and Handling

2° to 8°C