Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefoxitin, 98%

A broad spectrum antibiotic

Chemical Identifiers

Chemical Name or Material Cefoxitin
CAS 35607-66-0
Formula Weight 427.45
MDL Number MFCD00072014
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6689103

  SDS
alfa aesar™
J6689103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 427.45
Melting Point 149° to 150°C
Chemical Name or Material Cefoxitin
CAS 35607-66-0
Health Hazard 1 Warning
RTECS Number XI0386500
Physical Form Powder
Assay 98%
MDL Number MFCD00072014
Solubility Information Predicted solubility in water is less than 0.2mg/mL
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 843°C
Color White to off-white
Storage Note 1 2° to 8°C
Quantity 1g

Applications
Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfering with cell wall synthesis. Its activity spectrum includes a broad range of gram-negative and gram-positive bacteria including anaerobes.

Solubility
Predicted solubility in water is less than 0.2mg/ml

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.

2° to 8°C