Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefotiam, 98%


Quantity 100mg

Manufacturer: alfa aesar™  J66894MC

 View more versions of this product

Catalog No. AAJ66894MC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 525.63
Chemical Name or Material Cefotiam
MDL Number MFCD00865087
CAS 61622-34-2
Physical Form Powder
Assay 98%
Solubility Information Soluble in water
Boiling Point 843°C
Color White
Storage Note 1 2° to 8°C
Quantity 100mg
RTECS Number XI0366000

Specifications

Chemical Name or Material Cefotiam
CAS 61622-34-2
Formula Weight 525.63
MDL Number MFCD00865087

  • Interferes with PBP (penicillin binding protein) activity involved in the final phase of peptidoglycan synthesis

Accessories

2° to 8°C