Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefotiam, 98%

Chemical Identifiers

Chemical Name or Material Cefotiam
CAS 61622-34-2
Formula Weight 525.63
MDL Number MFCD00865087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66894MC

 
alfa aesar™
J66894MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66894ME

 
alfa aesar™
J66894ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 525.63
Chemical Name or Material Cefotiam
MDL Number MFCD00865087
CAS 61622-34-2
Physical Form Powder
Assay 98%
Solubility Information Soluble in water
Boiling Point 843°C
Color White
Storage Note 1 2° to 8°C
Quantity 100mg
RTECS Number XI0366000

  • Interferes with PBP (penicillin binding protein) activity involved in the final phase of peptidoglycan synthesis

Safety and Handling

2° to 8°C