Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefotiam, 98%


Quantity 500mg

Manufacturer: alfa aesar™  J66894ME

 View more versions of this product

Catalog No. AAJ66894ME

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 525.63
Assay 98%
MDL Number MFCD00865087
Chemical Name or Material Cefotiam
Physical Form Powder
CAS 61622-34-2
Solubility Information Soluble in water
Boiling Point 843°C
Color White
Storage Note 1 2° to 8°C
Quantity 500mg
RTECS Number XI0366000

Specifications

Chemical Name or Material Cefotiam
CAS 61622-34-2
Formula Weight 525.63
MDL Number MFCD00865087

  • Interferes with PBP (penicillin binding protein) activity involved in the final phase of peptidoglycan synthesis

Accessories

2° to 8°C