missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefotaxime sodium salt, 50mg/mL in distilled water, sterile-filtered

$32.32 - $32.32

Chemical Identifiers

CAS 64485-93-4
MDL Number MFCD00079073
Chemical Name or Material Cefotaxime sodium salt, 50mg/mL in distilled water, sterile-filtered
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ670058EQ SDS Alfa Aesar™
J670058EQ
5 x 1mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67005XF SDS Alfa Aesar™
J67005XF
1mL Each for $32.32
Specifications Description & Specifications

Chemical Identifiers

64485-93-4
MFCD00079073
Cefotaxime sodium salt, 50mg/mL in distilled water, sterile-filtered

Specifications

64485-93-4
MFCD00079073
Cefotaxime sodium salt, 50mg/mL in distilled water, sterile-filtered
Liquid
5 x 1mL

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0250000

storageNote1 : -30° to -10°C