missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefotaxime sodium salt

$126.14 - $760.27

Chemical Identifiers

CAS 64485-93-4
Molecular Formula C16H16N5NaO7S2
Molecular Weight (g/mol) 477.442
MDL Number MFCD00079073
InChI Key AZZMGZXNTDTSME-LOSLGVLSSA-M
PubChem CID 88631411
IUPAC Name sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)[O-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6269003 View Documents Alfa Aesar™
J6269003
1g Each for $126.14
 
AAJ6269006 View Documents Alfa Aesar™
J6269006
5g Each for $262.16
 
AAJ6269014 View Documents Alfa Aesar™
J6269014
25g Each for $760.27
 
Description

Description

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Incompatible with strong oxidizing agents. Store in a cool place. Light sensitive.
Specifications

Chemical Identifiers

64485-93-4
477.442
AZZMGZXNTDTSME-LOSLGVLSSA-M
sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
C16H16N5NaO7S2
MFCD00079073
88631411
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)[O-].[Na+]

Specifications

64485-93-4
MFCD00079073
14,1933
AZZMGZXNTDTSME-LOSLGVLSSA-M
sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
88631411
Powder
1g
C16H16N5NaO7S2
5711411
Freely soluble in water
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)[O-].[Na+]
477.442
477.4
White to Yellow
Cefotaxime sodium salt
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0250000