Learn More
Cefepime hydrochloride monohydrate, Thermo Scientific Chemicals
Cefepime hydrochloride monohydrate, CAS # 123171-59-5, is a hydrochloride salt of the antibiotic derived from Acremonium fungus.
Supplier: Thermo Scientific Chemicals J6623706
Description
Cefepime hydrochloride monohydrate is used as a broad spectrum antibiotic targeting a wide variety of naturally antibiotic resistant Gram-positive and Gram-negative bacteria. It is resistant to hydrolysis by common plasmid and/or chromosomally-mediated β-lactamases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Cefepime hydrochloride is a fourth-generation cephalosporin antibiotic that works against most antibiotic resistant strains of bacteria
• Cefepime hydrochloride is a penicillin binding protein inhibitor that prevents peptidoglycan cross-linkage and leads to cell wall weakening and lysis
Application
• Cefepime hydrochloride monohydrate can be used for the laboratory study of microbial physiology and antibiotic resistance
• Cefepime has broad-spectrum activity; it is appropriate for research on bacteria responsible for resistant infections and physiological studies
Specifications
Cefepime Hydrochloride Monohydrate | |
Powder | |
150°C | |
MFCD01687679 | |
LRAJHPGSGBRUJN-OMIVUECESA-O | |
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride | |
129893984 | |
White to Yellow | |
Light sensitive |
Cefepime hydrochloride monohydrate | |
123171-59-5 | |
C19H29Cl2N6O6S2+ | |
Soluble in DMSO | |
C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl | |
572.497 | |
571.5 | |
5 g |
Chemical Identifiers
123171-59-5 | |
572.497 | |
LRAJHPGSGBRUJN-OMIVUECESA-O | |
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride |
C19H29Cl2N6O6S2+ | |
MFCD01687679 | |
129893984 | |
C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only