Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefazolin sodium salt, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Cefazolin sodium salt
CAS 27164-46-1
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00056883
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67012XF

  SDS
alfa aesar™
J67012XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

CAS 27164-46-1
MDL Number MFCD00056883
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Physical Form Liquid
RTECS Number XI0390000
Storage Note 1 -30° to -10°C
Quantity 1mL
Name Note 50mg/mL in distilled water, sterile-filtered
Health Hazard 1 Danger
Chemical Name or Material Cefazolin sodium salt
Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

-30° to -10°C