Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefamandole nafate

A second generation cephalosporin antibiotic

Chemical Identifiers

Chemical Name or Material Cefamandole nafate
CAS 42540-40-9
Formula Weight 513.5
MDL Number MFCD00864877
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6673506

  SDS
alfa aesar™
J6673506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ6673509

  SDS
alfa aesar™
J6673509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 513.5
Chemical Name or Material Cefamandole nafate
Health Hazard 2 GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00864877
Physical Form Powder
Melting Point 190°C
CAS 42540-40-9
Health Hazard 1 Danger
Solubility Information Freely soluble in water; Sparingly soluble in methanol
Storage Note 1 2° to 8°C
Color White
Quantity 5g
RTECS Number XI0380000
Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

2° to 8°C