missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cedarwood Oil (Histological), Fisher Chemical

Histologic clearing agent

Manufacturer:  Thermo Scientific O40500

 View more versions of this product

Catalog No. O40-500




Specifications

Specifications

Cedarwood Oil
8000-27-9
Pass Test
Glass Bottle
1.502 to 1.507 (at 20°C)
Soluble
0.948 to 0.970 (at 25°C)
Liquid
279°C
500mL
0.95g/cm³
0.29 OD max.
MFCD00132766
Pass Test (in ethanol)
Soluble
0.15% max.
Histological
Yellow

Chemical Identifiers

8000-27-9
MFCD00132766
Safety and Handling

Safety and Handling

SDS Documents