Please login to your online account to display your discounted pricing

Cedarwood Oil (Histological), Fisher Chemical

Histologic clearing agent

$357.08 - $357.08

Chemical Identifiers

Chemical Name or Material Cedarwood Oil
CAS 8000-27-9
Purity Grade Histological Grade
MDL Number MFCD00132766
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

O40-500

  SDS
fisher scientific company
O40500
Glass Bottle 500mL Each for $357.08
Case of 6 Each for $2,142.48
Description & Specifications

Specifications

Purity Grade Histological Grade
Boiling Point 279°C
Quantity 500mL
Solubility in 2_Propanol Soluble
Water 0.15% max.
Chemical Name or Material Cedarwood Oil
Physical Form Liquid
MDL Number MFCD00132766
CAS 8000-27-9
Density 0.95g/cm³
Identification Pass Test
Optical Density 0.29 OD max.
Packaging Glass Bottle
Refractive Index 1.502 to 1.507 (at 20°C)
ChemAlert Storage Symbol Gray
Solubility Pass Test (in ethanol)
Solubility in Methanol Soluble
Specific Gravity 0.948 to 0.970 (at 25°C)
Viscosity [NULL]
Color Light Yellow
Safety and Handling