missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Cedarwood oil

 

Manufacturer:  Thermo Scientific™ 612085000

Catalog No. AC612085000



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Specifications

Specifications

500mL
8000-27-9
0.29 max.
MFCD00132766
Solubility in water: insoluble
SOLUBLE SOLUBILITY
0.15% max.
Cedarwood oil
Pass Test
EcoSafPak™
1.502 to 1.507
SOLUBLE SOLUBILITY
0.948 - 0.970

Chemical Identifiers

8000-27-9
MFCD00132766
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only