Please login to your online account to display your discounted pricing

Alfa Aesar™ Cecropin P1, Porcine

For cellular and molecular biology applications

$284.77 - $389.43

Specifications

Product Type Cecropin P1, porcine
Cross Reactivity Porcine
Molecular Weight 3338.85
Storage Requirements -30° to -10°C
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66344EXD

 
alfa aesar™
J66344EXD
0.5mg Each for $284.77

AAJ66344MCR

 
alfa aesar™
J66344MCR
1mg Each for $389.43
Description & Specifications

Specifications

Product Type Cecropin P1, porcine
Cross Reactivity Porcine
Molecular Weight 3338.85
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC