Please login to your online account to display your discounted pricing

Alfa Aesar™ Cecropin B, Free acid

For cellular and molecular biology applications

Specifications

Product Type Cecropin B, free acid
Molecular Weight 3834.66
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66041LB0

 
alfa aesar™
J66041LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66041MCR

 
alfa aesar™
J66041MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cecropin B, free acid
Molecular Weight 3834.66
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC