Please login to your online account to display your discounted pricing

Alfa Aesar™ Cecropin A (1-8)-Melittin (1-18)Amide

For cellular and molecular biology applications

Specifications

Product Type Cecropin A (1-8)-Melittin (1-18) amide
Molecular Weight 2794.50
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66814EXD

 
alfa aesar™
J66814EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66814MCR

 
alfa aesar™
J66814MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cecropin A (1-8)-Melittin (1-18) amide
Molecular Weight 2794.50
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-NH2
Regulatory Status RUO
Purification Method HPLC