Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS613P1ABX

 View more versions of this product

Catalog No. MS613P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Isotype IgG2a kappa
Clone COL-1
Regulatory Status RUO
Antigen CEA (Carcinoembryonic Antigen)/CD66e Ab-3
Cross Reactivity Human
Immunogen Human colon carcinoma extract
Host Species Mouse
Format Concentrated
Type Primary
Concentration 1mg/mL
Classification Monoclonal
Research Discipline Oncogenesis
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: COL-1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Immunogen: Human colon carcinoma extract

Molecular Weight: 180 kDa

Positive Control: LS174T cells, Colon CA

Cellular Localization: Cytoplasmic and luminal membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO