Invitrogen™ CDK9 Recombinant Human Protein, Inactive DFS Item
CDK9 is a member of the cyclin-dependent protein kinase family. CDK9 is closely related to cdc28 and cdc2 and is an important regulator of the cell cycle. Recombinant full-length human CDK9 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Manufacturer:  Invitrogen™ LSPV6305
 View more versions of this product
Catalog No. PV6305
Description
Description
CDK9 is a component of the multiprotein complex TAK/P-TEFβ. CDK9 can modulate RNA polymerase II-directed transcription by phosphorylating the C-terminal domain of the largest subunit of RNA polymerase II. CDK9 forms a complex with and is regulated by its regulatory subunit cyclin T or cyclin K. CDK9 also interacts with the HIV-1 Tat protein which suggested a possible involvement of this protein in AIDS.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
69.1kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Yes
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
100μg
CDC2L4, C-2k, TAK, PITALRE
Human
GST Tag
Documents