Invitrogen™ CDK9/Cyclin T1 Recombinant Human Protein DFS Item
CDK9 is coexpressed and copurified with CyclinT1.
Manufacturer:  Invitrogen™ LSPV4280
 View more versions of this product
Catalog No. PV4280
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
CDK9 is a serine/threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle.
Specifications
Specifications
NP_001252 I NP_001231
Frozen
131.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CDK and CHK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol
1mg
RUO - research use only
CDK9/Cyclin T1
Baculovirus
Human
His Tag
Documents