CDK7/Cyclin H/MNAT1 Recombinant Human Protein DFS Item
CDK7/Cyclin H/MNAT1 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4186
 View more versions of this product
Catalog No. PV4186
Description
Description
The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle.
Specifications
Specifications
Kinase assay
Baculovirus
0.4mg/mL
Frozen
126.3kDa
70% by SDS-PAGE
–80°C
1 Each
CDK7/Cyclin H/MNAT1 Recombinant Human Protein
CDK and CHK
96nmoles of phosphate transferred to CDK7/9tide peptide substrate (YSPTSPSYSPTSPSYSPTSPSKKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL
His Tag
NP_001790, Q130R, NP_001230, NP_002422
Human
Protein assays and analysis
Frozen
CDK7/Cyclin H/MNAT1 Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
CDK7/Cyclin H/MNAT1
CDK7/cyclin H/MNAT1 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 333μg/mL CDK7/9tide peptide substrate (YSPTSPSYSPTSPSYSPTSPSKKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
CDK7/cyclin H/MNAT1 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.