Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk6 Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

$268.31 - $1,256.61

Specifications

Antigen Cdk6 Ab-2
Clone K6.90; same as DCS-90
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS399P

 
thermo scientific™ lab vision™
MS399P
200μg/mL 1mL Each for $1,171.16

MS399P0

 
thermo scientific™ lab vision™
MS399P0
200μg/mL 100µL Each for $268.31

MS399P1

 
thermo scientific™ lab vision™
MS399P1
200μg/mL 500µL Each for $759.18

MS399P1ABX

 
thermo scientific™ lab vision™
MS399P1ABX
1mg/mL 100µL Each for $844.49

MS399PABX

 
thermo scientific™ lab vision™
MS399PABX
1mg/mL 200µL Each for $1,256.61
Description & Specifications

Specifications

Antigen Cdk6 Ab-2
Clone K6.90; same as DCS-90
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Purified recombinant cdk6 protein
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk6 Ab-2, Mouse Monoclonal Antibody.

Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: K6.90; same as DCS-90

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known

Epitope: Not determined

Immunogen: Purified recombinant Cdk6 protein

Molecular Weight: 40 kDa

Positive Control: Ls174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase assay (Not suitable)

Note:

USA: RUO; Int'l: RUO