Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk6 Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS399P1ABX

 View more versions of this product

Catalog No. MS399P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1
Format Concentrated
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Regulatory Status RUO
Antigen Cdk6 Ab-2
Quantity 100µL
Clone K6.90; same as DCS-90
Immunogen Purified recombinant cdk6 protein
Type Primary
Conjugate Unlabeled
Research Discipline Cell Cycle
Classification Monoclonal
Concentration 1mg/mL

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk6 Ab-2, Mouse Monoclonal Antibody.

Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: K6.90; same as DCS-90

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known

Epitope: Not determined

Immunogen: Purified recombinant Cdk6 protein

Molecular Weight: 40 kDa

Positive Control: Ls174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase assay (Not suitable)

Note:

USA: RUO; Int'l: RUO