missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk6 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

$278.04 - $1,531.02

Specifications

Antigen Cdk6 Ab-1
Clone K6.83; same as DCS-83
Applications Immunofluorescence
Classification Monoclonal
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS398B Thermo Scientific™ Lab Vision™
MS398B
200μg/mL Biotin 1mL Each for $1,531.02
MS398B0 Thermo Scientific™ Lab Vision™
MS398B0
200μg/mL Biotin 100μL Each for $372.86
MS398B1 Thermo Scientific™ Lab Vision™
MS398B1
200μg/mL Biotin 500μL Each for $1,014.31
MS398P Thermo Scientific
MS398P
200μg/mL Unlabeled 1mL Each for $1,176.31
MS398P0 Thermo Scientific™ Lab Vision™
MS398P0
200μg/mL Unlabeled 100μL Each for $278.04
MS398P1 Thermo Scientific™ Lab Vision™
MS398P1
200μg/mL Unlabeled 500μL Each for $778.55
MS398P1ABX Thermo Scientific™ Lab Vision™
MS398P1ABX
1mg/mL Unlabeled 100μL Each for $866.27
MS398PABX Thermo Scientific™ Lab Vision™
MS398PABX
1mg/mL Unlabeled 200μL Each for $1,302.88
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk6 Ab-1, Mouse Monoclonal Antibody.

Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks that are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: K6.83; same as DCS-83

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 30-65

Immunogen: Purified recombinant Cdk6 protein

Molecular Weight: 40 kDa

Positive Control: LS174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase assay (Not suitable)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cdk6 Ab-1
Immunofluorescence
Concentrated
RUO
Primary
IgG1
K6.83; same as DCS-83
Monoclonal
Murine
Cell Cycle
Purified recombinant cdk6 protein