Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk6 Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS398P1ABX

 View more versions of this product

Catalog No. MS398P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen Purified recombinant cdk6 protein
Isotype IgG1
Clone K6.83; same as DCS-83
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunofluorescence
Quantity 100µL
Format Concentrated
Antigen Cdk6 Ab-1
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk6 Ab-1, Mouse Monoclonal Antibody.

Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks that are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: K6.83; same as DCS-83

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 30-65

Immunogen: Purified recombinant Cdk6 protein

Molecular Weight: 40 kDa

Positive Control: LS174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase assay (Not suitable)

Note:

USA: RUO; Int'l: RUO