Invitrogen™ CDK5/p35 Recombinant Human Protein DFS Item
CDK5 is a cyclin-dependent kinase that plays a critical role in neuron development.
Manufacturer:  Invitrogen™ LSPV3981
 View more versions of this product
Catalog No. PV3981
Description
Description
The CDK5 kinase is strongly activated when it complexes with neuron-specific activators, especially p35, but less strongly when it complexes with cyclins. This protein is coexpressed and copurified with p35.
Specifications
Specifications
NP_004926.1 I NP_003876
Frozen
74.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CDK and CHK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 20% glycerol
1mg
RUO - research use only
CDK5/p35
Baculovirus
Human
His Tag
Documents