missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Human CDK5/p25, GST Tag, His Tag Recombinant Protein
Human CDK5/p25, GST Tag, His Tag Recombinant Protein
Supplier: Thermo Fisher Scientific PV4676
Description
CDK5 plays a critical role in neuron development. The disregulation of the CDK5 and the truncated activator p25 is believed to be involved in neurodegenerative diseases such as Alzheimer's, Parkinson's, and Amyotrophic Lateral Sclerosis. This protein is coexpressed and copurified with p25.Specifications
NP_004926.1 I NP_003876 | |
36.7⁄51.0 kDa | |
CDK5/p25, CDK5R | |
Human | |
GST Tag, His Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
CDK5 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
10 μg | |
CDK and CHK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMCG Group | |
Z'-LYTE™ | |
cyclin-dependent kinase 5 | |
Kinases & Inhibitors | |
Liquid | |
Cell Cycle & Proliferation, Signal Transduction |