Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk5 Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation applications with Thermo Scientific™ Cdk5 Ab-4 Mouse Monoclonal Antibody.

$309.24 - $1,381.06

Specifications

Antigen Cdk5 Ab-4
Clone DC17 + DC34
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1059P

 
thermo scientific™ lab vision™
MS1059P
200μg/mL 1mL Each for $1,279.96

MS1059P0

 
thermo scientific™ lab vision™
MS1059P0
200μg/mL 100μL Each for $309.24

MS1059P1

 
thermo scientific™ lab vision™
MS1059P1
200μg/mL 500μL Each for $804.92

MS1059P1ABX

 
thermo scientific™ lab vision™
MS1059P1ABX
1mg/mL 100μL Each for $875.29

MS1059PABX

 
thermo scientific™ lab vision™
MS1059PABX
1mg/mL 200μL Each for $1,381.06
Description & Specifications

Specifications

Antigen Cdk5 Ab-4
Clone DC17 + DC34
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Recombinant cdk5 protein
Isotype IgG1

Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation applications with Thermo Scientific™ Cdk5 Ab-4, Mouse Monoclonal Antibody.

Cdk5 is expressed in both cycling and noncycling cells and is present in numerous mammalian tissues. Neuronal and muscle cells contain the highest amount of this protein. The p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.

Host Species: Mouse

Clone: DC17 + DC34

Isotype: IgG1 + IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant Cdk5 protein

Molecular Weight: 31 kDa

Positive Control: IMR-5 or Ls174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO