Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk5 Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation applications with Thermo Scientific™ Cdk5 Ab-4 Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1059PABX

 View more versions of this product

Catalog No. MS1059PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cdk5 Ab-4
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone DC17 + DC34
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Immunogen Recombinant cdk5 protein
Isotype IgG1
Quantity 200μL
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human
Target Species Murine
Target Species Rat

Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation applications with Thermo Scientific™ Cdk5 Ab-4, Mouse Monoclonal Antibody.

Cdk5 is expressed in both cycling and noncycling cells and is present in numerous mammalian tissues. Neuronal and muscle cells contain the highest amount of this protein. The p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.

Host Species: Mouse

Clone: DC17 + DC34

Isotype: IgG1 + IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant Cdk5 protein

Molecular Weight: 31 kDa

Positive Control: IMR-5 or Ls174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO