Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk5 Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation applications with Thermo Scientific™ Cdk5 Ab-4 Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1059P1ABX

 View more versions of this product

Catalog No. MS1059P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Recombinant cdk5 protein
Clone DC17 + DC34
Host Species Mouse
Quantity 100µL
Format Concentrated
Isotype IgG1
Antigen Cdk5 Ab-4
Regulatory Status RUO
Classification Monoclonal
Concentration 1mg/mL
Research Discipline Cell Cycle
Conjugate Unlabeled
Type Primary

Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation applications with Thermo Scientific™ Cdk5 Ab-4, Mouse Monoclonal Antibody.

Cdk5 is expressed in both cycling and noncycling cells and is present in numerous mammalian tissues. Neuronal and muscle cells contain the highest amount of this protein. The p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.

Host Species: Mouse

Clone: DC17 + DC34

Isotype: IgG1 + IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant Cdk5 protein

Molecular Weight: 31 kDa

Positive Control: IMR-5 or Ls174T cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO