missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk5 Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk5 Ab-3, Mouse Monoclonal Antibody.

$522.65 - $1,597.02

Specifications

Antigen Cdk5 Ab-3
Clone DC34
Applications Immunofluorescence,Immunoprecipitation,Immunofluorescence,Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS906P Thermo Scientific™ Lab Vision™
MS906P
200μg/mL 1mL Each for $1,473.88
MS906P0 Thermo Scientific™ Lab Vision™
MS906P0
200μg/mL 100μL Each for $522.65
MS906P1 Thermo Scientific™ Lab Vision™
MS906P1
200μg/mL 500μL Each for $1,116.47
MS906P1ABX Thermo Scientific™ Lab Vision™
MS906P1ABX
1mg/mL 100μL Each for $1,239.61
MS906PABX Thermo Scientific™ Lab Vision™
MS906PABX
1mg/mL 200μL Each for $1,597.02
Specifications

Description

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk5 Ab-3, Mouse Monoclonal Antibody.

Cdk5 is expressed in both cycling and noncycling cells and is present in numerous mammalian tissues. Neuronal and muscle cells contain the highest amount of this protein. The p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.

Host Species: Mouse

Clone: DC34

Isotype: IgG1/κ

Species Reactivity: Human, Rat, Mouse and C. elegans. Others not known

Epitope: Not determined

Immunogen: Recombinant Cdk5 protein

Molecular Weight: 31 kDa

Positive Control: HT29, MAD109 or PC12 cells; Brain

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cdk5 Ab-3
Immunofluorescence,Immunoprecipitation,Immunofluorescence,Immunoprecipitation
Unlabeled
Murine
Cell Cycle
Recombinant cdk5 protein
DC34
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa