Learn More
Cdk5 Ab-3 Mouse Monoclonal Antibody, Epredia™
Cdk5 Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Thermo Fisher Scientific MS906P1
Description
Cdk5 is expressed in both cycling and noncycling cells and is present in numerous mammalian tissues. Neuronal and muscle cells contain the highest amount of this protein. The p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.Host Species: Mouse
Clone: DC34
Isotype: IgG1/κ
Species Reactivity: Human, Rat, Mouse and C. elegans. Others not known
Epitope: Not determined
Immunogen: Recombinant Cdk5 protein
Molecular Weight: 31 kDa
Positive Control: HT29, MAD109 or PC12 cells; Brain
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Cdk5 Ab-3 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Cell Cycle | |
C. elegans, Human, Mouse, Rat | |
IgG2a κ |
Immunofluorescence, Immunoprecipitation | |
DC34 | |
Unconjugated | |
Recombinant cdk5 protein | |
RUO | |
Primary | |
Concentrated |