Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk4 Ab-6, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk4 Ab-6, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS616P1ABX

 View more versions of this product

Catalog No. MS616P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Immunogen Purified recombinant cdk4 protein
Antigen Cdk4 Ab-6
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Quantity 100µL
Regulatory Status RUO
Clone DCS-31 + DCS-35
Conjugate Unlabeled
Concentration 1mg/mL
Type Primary
Classification Monoclonal
Research Discipline Cell Cycle

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk4 Ab-6, Mouse Monoclonal Antibody.

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-31 + DCS-35

Isotype: IgG1 + IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1-35

Immunogen: Purified recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: MCF-7 or PC12 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO