Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk4 Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk4 Ab-2, Mouse Monoclonal Antibody.

$257.92 - $1,180.51

Specifications

Antigen Cdk4 Ab-2
Clone DCS-31
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS469P

 
thermo scientific™ lab vision™
MS469P
200μg/mL 1mL Each for $1,111.39

MS469P0

 
thermo scientific™ lab vision™
MS469P0
200μg/mL 100µL Each for $257.92

MS469P1

 
thermo scientific™ lab vision™
MS469P1
200μg/mL 500µL Each for $721.27

MS469P1ABX

 
thermo scientific™ lab vision™
MS469P1ABX
1mg/mL 100µL Each for $796.35

MS469PABX

 
thermo scientific™ lab vision™
MS469PABX
1mg/mL 200µL Each for $1,180.51
Description & Specifications

Specifications

Antigen Cdk4 Ab-2
Clone DCS-31
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Recombinant cdk4 protein
Isotype IgG1

Obtain accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk4 Ab-2, Mouse Monoclonal Antibody.

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-31

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 16-35

Immunogen: Recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: MAD109, LS174T or PC12 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO