Invitrogen™ CDK2/cyclin A1 Recombinant Human Protein DFS Item
CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed. Recombinant full-length human CDK2 and CyclinA1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
$612.00 - $9,380.00
Specifications
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6289 DFS Item
View Documents
Invitrogen™
LSPV6289
10μg
Each for $612.00
PV6290 DFS Item
View Documents
Invitrogen™
LSPV6290
100μg
Each for $2,095.00
PV6291 DFS Item
View Documents
Invitrogen™
LSPV6291
1mg
Each for $9,380.00
Description
Description
CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. CDK2 associates with and is regulated by the regulatory subunits of the complex including Cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). CDK2 phosphorylates multiple cellular substrates including SMAD3 and FOXO1. Phosphorylation of FOXO1 leads to its inhibition.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
GST Tag
Yes
SDS