Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk2 Ab-4, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk2 Ab-4, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS617P1ABX

 View more versions of this product

Catalog No. MS617P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2a IgG2b
Host Species Mouse
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Clone 2B6 + 8D4
Antigen Cdk2 Ab-4
Immunogen Human recombinant cdk2 protein
Regulatory Status RUO
Format Concentrated
Research Discipline Cell Cycle
Type Primary
Conjugate Unlabeled
Concentration 1mg/mL
Classification Monoclonal

Ensure accurate, reproducible results in Western Blotting, Immunofluorescence and Immunoprecipitation experiments with Thermo Scientific™ Cdk2 Ab-4, Mouse Monoclonal Antibody.

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.

Host Species: Mouse

Clone: 2B6 + 8D4

Isotype: IgG2b (2B6) + IgG2a (8D4)

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant Cdk2 protein

Molecular Weight: 33 kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured only)

Note:

USA: RUO; Int'l: RUO