Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk2 Ab-1, Mouse Monoclonal Antibody


Accurately detect Cdk2 in human, mouse and rat tissues with Thermo Scientific™ Cdk2 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS459P1ABX

 View more versions of this product

Catalog No. MS459P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Antigen Cdk2 Ab-1
Format Concentrated
Host Species Mouse
Immunogen Human recombinant cdk2 protein
Isotype IgG2b
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 100µL
Clone 2B6
Type Primary
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cell Cycle
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Accurately detect Cdk2 in human, mouse and rat tissues with Thermo Scientific™ Cdk2 Ab-1, Mouse Monoclonal Antibody.

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as is cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.

Host Species: Mouse

Clone: 2B6

Isotype: IgG2b

Species Reactivity: Human, Mouse and Rat.

Epitope: Not determined

Immunogen: Human recombinant Cdk2 protein

Molecular Weight: 33 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO