Invitrogen™ CDK14 (PFTK1)/cyclin Y Recombinant Human Protein DFS Item
CDK14 (PFTK1)/cyclin Y is a member of the CDC2-related protein kinase family. Recombinant human full-length CDK14 and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.
$612.00 - $9,380.00
Specifications
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6382 DFS Item
View Documents
Invitrogen™
LSPV6382
10μg
Each for $612.00
PV6384 DFS Item
View Documents
Invitrogen™
LSPV6384
1mg
Each for $9,380.00
PV6383 DFS Item
View Documents
Invitrogen™
LSPV6383
100μg
Each for $2,095.00
Description
Description
CDK14 (PFTK1)/cyclin Y is expressed primarily in the postnatal and adult nervous system. CDK14 is expressed at high levels in brain, pancreas, kidney, heart, testis, and ovary. CDK14 amino acid residue ser119 is required for its interaction with all four 14-3-3 isoforms, while ser120 may contribute slightly to CDK14 binding. CDK14 acts as a cyclin-dependent kinase that regulates cell cycle progression and cell proliferation. Overexpression of CDK14 predicts resistance to chemotherapy in patients with oesophageal squamous cell carcinoma.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
GST Tag
Yes
SDS