Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdk1/p34 cdc2 Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and kinase assay applications. 

Manufacturer: thermo scientific™ lab vision™  MS110P1ABX

 View more versions of this product

Catalog No. MS110P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cdk1/p34superscriptcdc2superscript Ab-1
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Woodchuck
Cross Reactivity Xenopus
Format Concentrated
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli
Isotype IgG2a
Regulatory Status RUO
Quantity 100µL
Clone A17.1.1
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cell Cycle
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cdk1/p34cdc2 Ab-1, Mouse Monoclonal Antibody.

p34cdc2(a catalytic subunit of Maturation Promoting Factor) plays a crucial role during cell division and is most active during mitosis. It is a serine/threonine kinase, which is activated by cyclin through dephosphorylation of tyrosine residues. p34cdc2 is inactivated by a tyrosine kinase.

Host Species: Mouse

Clone: A17.1.1

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat, Guinea Pig, Woodchuck, Xenopus and Chicken.

Epitope: aa 220-227

Immunogen: C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli

Molecular Weight: 34 kDa

Positive Control: HeLa, MAD109, PC12 or NIH-3T3 cells; Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase Assay
  • Inhibits activation of p34cdc2 kinase by cyclins

Note:

USA: RUO; Int'l: RUO