Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cdh1 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Cdh1 Ab-1, Mouse Monoclonal Antibody.

$288.41 - $1,452.65

Specifications

Antigen Cdh1 Ab-1
Clone DH01; same as DCS-266
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1116P

 
thermo scientific
MS1116P
200μg/mL 1mL Each for $1,278.59

MS1116P0

 
thermo scientific™ lab vision™
MS1116P0
200μg/mL 100μL Each for $288.41

MS1116P1

 
thermo scientific™ lab vision™
MS1116P1
200μg/mL 500μL Each for $876.55

MS1116P1ABX

 
thermo scientific™ lab vision™
MS1116P1ABX
1mg/mL 100μL Each for $882.73

MS1116PABX

 
thermo scientific™ lab vision™
MS1116PABX
1mg/mL 200μL Each for $1,452.65
Description & Specifications

Specifications

Antigen Cdh1 Ab-1
Clone DH01; same as DCS-266
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Recombinant human cdh1 protein
Isotype IgG2a kappa

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Cdh1 Ab-1, Mouse Monoclonal Antibody.

Research Areas: Cell Cycle; Cdh1 protein is a limiting, substrate-specific activator of anaphase-promoting complex (APC)-dependent proteolysis and it is required for the degradation of the APC substrates, such as Clb2 and Ase1. Cdh1 plays important role in mitotic regulation.

Host Species: Mouse

Clone: DH01; same as DCS-266

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human Cdh1 protein

Molecular Weight: ∽55 kDa

Positive Control: Jurkat or LS174T cells

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO