Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CDC7 Kinase Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS1888PABX

 View more versions of this product

Catalog No. MS1888PABX

  • / Each
 

Description & Specifications

Specifications

Immunogen GST fusion protein full length CDC7
Regulatory Status RUO
Isotype IgG1
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Concentrated
Quantity 200µL
Clone DCS-341
Host Species Mouse
Cross Reactivity Human
Antigen CDC7 Kinase Ab-1
Type Primary
Conjugate Unlabeled
Research Discipline Cell Cycle
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ CDC7 Kinase Ab-1, Mouse Monoclonal Antibody.

CDC7, a catalytic subunit of Dbf4 is an important regulator of S-phase in human cells.

Host Species: Mouse

Clone: DCS-341

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: GST fusion protein full length CDC7

Positive Control: HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO