Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CDC7 Kinase Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS1888P1ABX

 View more versions of this product

Catalog No. MS1888P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen CDC7 Kinase Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone DCS-341
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen GST fusion protein full length CDC7
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ CDC7 Kinase Ab-1, Mouse Monoclonal Antibody.

CDC7, a catalytic subunit of Dbf4 is an important regulator of S-phase in human cells.

Host Species: Mouse

Clone: DCS-341

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: GST fusion protein full length CDC7

Positive Control: HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO