Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CDC47/MCM7 Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS862P1ABX

 View more versions of this product

Catalog No. MS862P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Format Concentrated
Antigen CDC47/MCM7 Ab-2
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Immunogen Recombinant hCDC47 protein
Clone 47DC141; same as DCS-141
Quantity 100µL
Host Species Mouse
Classification Monoclonal
Research Discipline Cell Cycle
Type Primary
Conjugate Unlabeled
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)

Obtain accurate, reproducible results in Immunohistochemistry, Western Blotting, Immunofluorescence and Immunoprecipitation experiments with Thermo Scientific™ CDC47/MCM7 Ab-2, Mouse Monoclonal Antibody.

hCDC47 is a human member of the MCM family, which allows DNA to replicate once per cell cycle. In quiescent cells, human MCM7 (hMCM7) mRNA is almost undetectable. Stimulation of cells to enter the cell cycle results in induction of hMCM7 expression. The hCDC47 protein expression and localization is found in nuclei of the proliferative components of normal lymph nodes, bone marrow, epidermis and mucosa. Malignant tumors from several organs contained more nuclear hCDC47-positive cells than their normal counterparts. These results indicate that hCDC47 may play a role in normal and neoplastic cell growth in vivo.

Host Species: Mouse

Clone: 47DC141; same as DCS-141

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Dog and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Recombinant hCDC47 protein

Molecular Weight: 80 kDa

Positive Control: MAD109, or PC12 cells, Tonsil or breast cancer

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO