CDC42BPB (MRCKB) Recombinant Human Protein DFS Item
CDC42 BPB (MRCKB)
Manufacturer:  Life Technologies LSPV4399
Catalog No. PV4399
Description
Description
CDC42BPB is a serine⁄threonine kinases which functions in actin cytoskeleton assembly and organization.
Specifications
Specifications
NP_006026.2
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
CDC42BPB (MRCKB) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
CDC42BPB (MRCKB) Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 1-473
279nmoles of phosphate transferred to S6 peptide substrate (AKRRRLSSLRA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL
His Tag
0.47mg/mL
58.9kDa
90% by SDS-PAGE
–80°C
1 Each
CDC42BPB (MRCKB)
CDC42 BPB (MRCKB) was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL S6 peptide substrate (AKRRRLSSLRA) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
DMPK
CDC42BPB (MRCKB) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.