Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CDC25C Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting, immunofluorescence, and immunoprecipitation experiments.

$458.59 - $1,563.29

Specifications

Antigen CDC25C Ab-1
Clone 25C14; same as TC-14
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS751P

 
thermo scientific™ lab vision™
MS751P
200μg/mL 1mL Each for $1,388.90

MS751P0

 
thermo scientific™ lab vision™
MS751P0
200μg/mL 100μL Each for $458.59

MS751P1

 
thermo scientific™ lab vision™
MS751P1
200μg/mL 500μL Each for $1,081.04

MS751P1ABX

 
thermo scientific™ lab vision™
MS751P1ABX
1mg/mL 100μL Each for $1,213.43

MS751PABX

 
thermo scientific™ lab vision™
MS751PABX
1mg/mL 200μL Each for $1,563.29
Description & Specifications

Specifications

Antigen CDC25C Ab-1
Clone 25C14; same as TC-14
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Recombinant human CDC25C protein
Isotype IgG2a kappa

Obtain accurate, reproducible results in western blotting, immunofluorescence, and immunoprecipitation experiments with Thermo Scientific™ CDC25C Ab-1, Mouse Monoclonal Antibody.

Cyclin-dependent kinases can be activated by CDC25, which removes inhibitory phosphates from tyrosine and threonine residues. At least three CDC25 genes (CDC25A, CDC25B, and CDC25C) have been identified in humans. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Host Species: Mouse

Clone: 25C14; same as TC-14

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: aa 1-150

Immunogen: Recombinant human CDC25C protein

Molecular Weight: 53-56 kDa

Positive Control: HeLa, HT29 or LS174T cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO