Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CDC25C Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting, immunofluorescence, and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS751P1ABX

 View more versions of this product

Catalog No. MS751P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen Recombinant human CDC25C protein
Cross Reactivity Human
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Clone 25C14; same as TC-14
Antigen CDC25C Ab-1
Format Concentrated
Isotype IgG2a kappa
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Obtain accurate, reproducible results in western blotting, immunofluorescence, and immunoprecipitation experiments with Thermo Scientific™ CDC25C Ab-1, Mouse Monoclonal Antibody.

Cyclin-dependent kinases can be activated by CDC25, which removes inhibitory phosphates from tyrosine and threonine residues. At least three CDC25 genes (CDC25A, CDC25B, and CDC25C) have been identified in humans. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Host Species: Mouse

Clone: 25C14; same as TC-14

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: aa 1-150

Immunogen: Recombinant human CDC25C protein

Molecular Weight: 53-56 kDa

Positive Control: HeLa, HT29 or LS174T cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO