missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CDC25A Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ CDC25A Ab-1, Mouse Monoclonal Antibody.

$277.12 - $1,296.43

Specifications

Antigen CDC25A Ab-1
Clone DCS-120
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS638P Thermo Scientific™ Lab Vision™
MS638P
200μg/mL 1mL Each for $1,182.35
MS638P0 Thermo Scientific™ Lab Vision™
MS638P0
200μg/mL 100μL Each for $277.12
MS638P1 Thermo Scientific™ Lab Vision™
MS638P1
200μg/mL 500μL Each for $776.16
MS638PABX Thermo Scientific™ Lab Vision™
MS638PABX
1mg/mL 200μL Each for $1,296.43
MS638P1ABX Thermo Scientific
MS638P1ABX
1mg/mL 100μL Each for $849.80
Specifications

Description

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ CDC25A Ab-1, Mouse Monoclonal Antibody.

Cyclin-dependent kinases are positively regulated by association with cyclins and negatively regulated by binding to inhibitory subunits. The activity of cyclin-dependent kinases is also regulated by the phosphorylation status, which is controlled by the antagonistic action of wee1 kinase and CDC25 phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Host Species: Mouse

Clone: DCS-120

Isotype: IgG2a

Species Reactivity: Human and Rat. Others not known.

Immunogen: Purified recombinant CDC25A

Molecular Weight: 65 kDa

Positive Control: LS174T cells, Tonsil

Cellular Localization: Nuclear membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CDC25A Ab-1
Immunoprecipitation
Unlabeled
Murine
Cell Cycle
Purified recombinant CDC25A
DCS-120
Monoclonal
Human,Rat,Human,Rat
RUO
Primary
IgG2a