Please login to your online account to display your discounted pricing

CD99 Mouse anti-Human, PE, Clone: 3B2/TA8, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen CD99
Clone 3B2/TA8
Host Species Mouse
Gene Alias E2 antigen, MIC2, MIC 2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010535

 
life technologies
LS12099741
25 tests Each for $143.55

5010536

 
life technologies
LS12099742
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen CD99
Clone 3B2/TA8
Host Species Mouse
Gene Alias E2 antigen, MIC2, MIC 2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The 3B2/TA8 monoclonal antibody reacts with human CD99, also known as E2 antigen and MIC2. CD99 is an approximately 32kDa sialoglycoprotein and is expressed highly by all leukocytes. However, there is little to no expression in granulocytes. CD99 is involved in T cell adhesion and activation.