Please login to your online account to display your discounted pricing

CD99 Mouse anti-Human, FITC, Clone: 3B2/TA8, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $288.55

Specifications

Antigen CD99
Clone 3B2/TA8
Host Species Murine
Gene Alias E2 antigen, MIC2, MIC 2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509716

 
life technologies
LS11099741
25 tests Each for $100.05

509717

 
life technologies
LS11099742
100 Tests Each for $288.55
Description & Specifications

Specifications

Antigen CD99
Clone 3B2/TA8
Host Species Murine
Gene Alias E2 antigen, MIC2, MIC 2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The 3B2/TA8 monoclonal antibody reacts with human CD99, also known as E2 antigen and MIC2. CD99 is an approximately 32kDa sialoglycoprotein and is expressed by all leukocytes. CD99 is involved in T cell adhesion and activation.