Please login to your online account to display your discounted pricing

CD98hc Mouse anti-Human, FITC, Clone: 5E5, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD98hc
Clone 5.00E+05
Host Species Mouse
Gene Alias 4F2hc, SLC3A2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122084

 
life technologies
LS11098241
25 tests Each for $143.55

501122085

 
life technologies
LS11098242
100 tests Each for $317.55
Description & Specifications

Specifications

Antigen CD98hc
Clone 5.00E+05
Host Species Mouse
Gene Alias 4F2hc, SLC3A2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The monoclonal antibody 5E5 recognizes human CD98hc protein also called CD98 heavy chain, FRP-1 (fusion regulatory protein-1 ) or 4F2. CD98 is a disulfide-linked complex composed of a heavy chain (a Type II membrane protein) and a light chain of which there are more than six different chains (LAT1, LAT2, y+LAT1, y+LAT2, Asc-1, xCT). CD98 can associate directly or indirectly with β 1 integrins, EpCAM and CD147 (EMMPRIN). The function of CD98 is thought to involve amino acid transport, cell fusion concomitantly with increased plasma membrane expression and integrin mediated adhesion. Expression of CD98 is broad including non-hematopoietic cells. In peripheral blood cells, monocytes expression is high while lymphocytes have intermediate levels and granulocytes express very little.